Skip to Content
Merck
All Photos(1)

Key Documents

52276

Supelco

1-Hexadecene

analytical standard

Synonym(s):

Cetene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$263.00

$263.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$263.00

About This Item

Linear Formula:
CH3(CH2)13CH=CH2
CAS Number:
Molecular Weight:
224.43
Beilstein:
1753401
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$263.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

>0.8 (vs air)

vapor pressure

<0.2 mmHg ( 20 °C)

Assay

≥99.8% (GC)

autoignition temp.

464 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.441 (lit.)
n20/D 1.441

bp

274 °C (lit.)

mp

3-5 °C (lit.)

density

0.783 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCC=C

InChI

1S/C16H32/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3H,1,4-16H2,2H3

InChI key

GQEZCXVZFLOKMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Contact allergy to polyvinylpyrrolidone/hexadecene copolymer previously reported.
A C De Groot
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 7(4), 260-260 (1996-12-01)
M Tobler et al.
Contact dermatitis, 26(5), 299-303 (1992-05-01)
In continuation of our preceding studies, we disclose long-term experiments to test more extensively the protective power of the novel 4H Glove. 2 compounds encountered in modern electron microscopy technique, 1-hexadecene (1-HD) and 2-hydroxyethyl acrylate (2-HEA), which elicit allergic and/or
C Barba et al.
Meat science, 90(3), 697-700 (2011-11-22)
Solid phase microextraction (SPME) coupled with either gas chromatography-ionization flame detector (CG-FID) or multidimensional gas chromatography-mass spectrometry (MDGC-MS) was evaluated for its ability to detect volatile hydrocarbons produced during the irradiation of cooked ham. The chromatogram of an irradiated sample
Vincent Grossi et al.
Applied and environmental microbiology, 81(9), 3157-3168 (2015-03-01)
Bacterial glycerol ether lipids (alkylglycerols) have received increasing attention during the last decades, notably due to their potential role in cell resistance or adaptation to adverse environmental conditions. Major uncertainties remain, however, regarding the origin, biosynthesis, and modes of formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service