Skip to Content
Merck
All Photos(2)

Key Documents

44148

Sigma-Aldrich

1-Dodecene

≥99.0% (GC)

Synonym(s):

n-Dodec-1-ene, alpha-Dodecene

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$112.00
50 ML
$391.00

$112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$112.00
50 ML
$391.00

About This Item

Linear Formula:
CH3(CH2)9CH=CH2
CAS Number:
Molecular Weight:
168.32
Beilstein:
1699848
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$112.00


Check Cart for Availability

Request a Bulk Order

vapor density

5.8 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

491 °F

expl. lim.

~4.7 %

refractive index

n20/D 1.429 (lit.)
n20/D 1.430

bp

214-216 °C (lit.)

mp

−35 °C (lit.)

density

0.758 g/mL at 20 °C (lit.)

functional group

allyl

SMILES string

CCCCCCCCCCC=C

InChI

1S/C12H24/c1-3-5-7-9-11-12-10-8-6-4-2/h3H,1,4-12H2,2H3

InChI key

CRSBERNSMYQZNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Degradation of 1-dodecene by Pseudomonas strain H has been reported.[1] Its hydroformylation by employing microemulsion as a reaction media has been studied.[2]

Application

1-Dodecene may be employed to compose the collection liquid for the stabilization of the newly synthesized stable, alkyl-terminated, light-emitting silicon nanoparticles.[3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F - No data available

Flash Point(C)

79 °C - No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alkylation of 1-dodecene with benzene over H3PW12O40 supported on mesoporous silica SBA-15.
Wang J and Zhu H-O.
Catalysis Letters, 93(3-4), 209-212 (2004)
Hydroformylation of 1-dodecene using Rh-TPPTS in a microemulsion.
Haumann M, et al.
Applied Catalysis A: General, 225(1), 239-249 (2002)
A Malumbres et al.
Nanoscale, 7(18), 8566-8573 (2015-04-22)
Stable, alkyl-terminated, light-emitting silicon nanoparticles have been synthesized in a continuous process by laser pyrolysis of a liquid trialkyl-silane precursor selected as a safer alternative to gas silane (SiH4). Stabilization was achieved by in situ reaction using a liquid collection
Martin Lommatzsch et al.
Journal of chromatography. A, 1402, 94-101 (2015-06-04)
Multidimensional chromatography based on two-dimensional high performance liquid chromatography on-line coupled to gas chromatography (on-line HPLC-HPLC-GC) enables the separate analysis of saturated, monounsaturated and aromatic hydrocarbons in packaging materials like polyolefins or paperboard and their migrates into foods. Since normal-phase
Melina Nisenbaum et al.
Journal of environmental sciences (China), 25(3), 613-625 (2013-08-08)
We report on the biodegradation of pure hydrocarbons and chemotaxis towards these compounds by an isolated chlorophenol degrader, Pseudomonas strain H. The biochemical and phylogenetic analysis of the 16S rDNA sequence identified Pseudomonas strain H as having 99.56% similarity with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service