Skip to Content
Merck
All Photos(1)

Key Documents

8.22064

Sigma-Aldrich

1-Hexadecene

for synthesis

Synonym(s):

1-Hexadecene, 1-Hexadecylene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$46.70
500 ML
$62.10

$46.70


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$46.70
500 ML
$62.10

About This Item

Linear Formula:
CH3(CH2)13CH=CH2
CAS Number:
Molecular Weight:
224.43
MDL number:
UNSPSC Code:
12352002
EC Index Number:
211-105-8
NACRES:
NA.22

$46.70


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

0.35 hPa ( 25 °C)

Quality Level

Assay

≥92.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.5-5.8 % (v/v)

kinematic viscosity

4.0 cSt(20 °C)

bp

274 °C/1013 hPa

mp

2.2 °C

transition temp

flash point 132 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCC)CCCCCCC=C

InChI

1S/C16H32/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3H,1,4-16H2,2H3

InChI key

GQEZCXVZFLOKMC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 92.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.780 - 0.789
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F

Flash Point(C)

132 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service