Skip to Content
Merck
All Photos(1)

Key Documents

51803

Millipore

Gruft Mycobacterial Supplement

suitable for microbiology

Synonym(s):

Mycobacterial Supplement according to Gruft

Sign Into View Organizational & Contract Pricing

Select a Size

5 AMP
$79.00

$79.00


Estimated to ship on31 May 2025


Request more information

Select a Size

Change View
5 AMP
$79.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

$79.00


Estimated to ship on31 May 2025


Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages
life science and biopharma
veterinary

microbiology

storage temp.

2-8°C

suitability

mycobacteria

General description

An antibiotic and enrichment supplement recommended for the selective cultivation of Mycobacteria. Added to TB-Medium according to Lowenstein Jensen (Cat. No.63237) penicillin and nalidixic acid inhibit bacteria Gram negative and Gram positive. Ribonucleic Acid is used as growth factor.

Application

for the selective cultivation of Mycobacteria

Components

(per vial sufficient for 400 mL medium)
Penicillin 20,000 IU, Nalidixic acid 14.0 mg, Ribonucleic acid 20.0 mg

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service