Skip to Content
Merck
All Photos(1)

Key Documents

34130

Supelco

Fusarenon X solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

3α,7α,15-Trihydroxy-4β-acetoxy-12,13-epoxytrichothec-9-en-8-one, FusX

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$731.00

$731.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$731.00

About This Item

Empirical Formula (Hill Notation):
C17H22O8
CAS Number:
Molecular Weight:
354.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$731.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1[C@@H](O)[C@H]2O[C@@H]3C=C(C)C(=O)[C@@H](O)[C@]3(CO)[C@]1(C)[C@]24CO4

InChI

1S/C17H22O8/c1-7-4-9-16(5-18,12(22)10(7)20)15(3)13(24-8(2)19)11(21)14(25-9)17(15)6-23-17/h4,9,11-14,18,21-22H,5-6H2,1-3H3/t9-,11-,12-,13-,14-,15-,16-,17+/m1/s1

InChI key

XGCUCFKWVIWWNW-CAYGJDLQSA-N

General description

Certan Vial
Fusarenon X, is a trichothecene mycotoxin[1], which is produced by Fusarium nivale fungi[2], naturally occurring in crops such as wheat and barley.[3]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low tumor incidence in rats with long-term feeding of fusarenon-X, a cytotoxic trichothecene produced by Fusarium nivale
Saito M, et al.
The Japanese Journal of Experimental Medicine, 50, 293-302 (1980)
The fates of trichothecene mycotoxins, nivalenol and fusarenon-X, in mice
Poapolathep A, et al.
Toxicon, 41, 1047-1054 (2003)
Fusarenon-X induced apoptosis in HL-60 cells depends on caspase activation and cytochrome c release
Miura K, et al.
Toxicology, 172, 103-112 (2002)
J Li et al.
The Journal of veterinary medical science, 59(3), 191-199 (1997-03-01)
Male 5-week-old Wistar rats orally (po) administered with fusarenon-X (FX) 1.5 mg/kg and control rats po-treated with distilled water were sacrificed at 0-48 hr after gavage. FX-administered rats showed significant dilatation of the stomach with increased fluid contents at 1-24
Margit Schollenberger et al.
International journal of food microbiology, 72(1-2), 85-89 (2002-02-15)
A total of 60 samples of wheat flour were collected during the first 6 months of 1999 from mills and food stores in an area in southwest Germany. Samples included whole-grain and two types of white flour with these three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service