Skip to Content
Merck
All Photos(1)

Key Documents

SML1664

Sigma-Aldrich

Deoxynivalenol Ready Made Solution

1 mg/mL

Synonym(s):

(3α,7α)-3,7,15-trihydroxy-12,13-epoxytrichothec-9-en-8-one, 4-Deoxynivalenol, DON, Dehydronivalenol, RD-toxon, Vomitoxin

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$209.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$209.00

About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
UNSPSC Code:
12352200
NACRES:
NA.77

$209.00


Check Cart for Availability

Request a Bulk Order

biological source

Fusarium sp. (Fusarium asiaticum)

Quality Level

form

solution (ethanol: 2-propanol 95:5)

storage condition

protect from light

concentration

1 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@]2([C@@H]3O[C@H]4[C@@]([C@]2(C[C@H]3O)C)([C@@H](C(=O)C(=C4)C)O)CO)C1

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Biochem/physiol Actions

Deoxynivalenol (DON) is a trichothecene mycotoxin that inhibits the synthesis of DNA and RNA as well as protein synthesis at the ribosomal level.
Deoxynivalenol (DON) is a trichothecene mycotoxin that inhibits the synthesis of DNA and RNA as well as protein synthesis at the ribosomal level. DON induces IL-6 mediated serum hyperelevation of IgA, as well as phosphorylation of extracellular signal regulated protein kinases 1 and 2 (ERK 1,2) and c-Jun N-terminal kinases 1 and 2 (JNK 1,2) in mice. An in vitro study with porcine ovarian granulosa cells suggests a dose dependent association of DON on porcine ovarian functions. It was also shown that LPS and its downstream mediators can interact with DON to modulate proliferative, cytotoxic and apoptotic outcomes in leukocytes in a tissue specific manner.

Preparation Note

Deoxynivalenol ready-made solution (1 mg/ml in Ethanol: 2-propanol 95:5) can be diluted in aqueous buffers to a working concentration of 0.1-1 μg/ml (1:1000).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service