Skip to Content
Merck
All Photos(1)

Key Documents

34131

Supelco

Nivalenol solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

3α,4β,7α, 15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one, NIV

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$840.00

$840.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$840.00

About This Item

Empirical Formula (Hill Notation):
C15H20O7
CAS Number:
Molecular Weight:
312.32
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$840.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)[C@@H](O)[C@@](C)([C@]34CO4)[C@@]1(CO)[C@H](O)C(=O)C(C)=C2

InChI

1S/C15H20O7/c1-6-3-7-14(4-16,11(20)8(6)17)13(2)10(19)9(18)12(22-7)15(13)5-21-15/h3,7,9-12,16,18-20H,4-5H2,1-2H3/t7-,9-,10-,11-,12-,13-,14-,15+/m1/s1

InChI key

UKOTXHQERFPCBU-XBXCNEFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial
Nivalenol is a potential trichothecene mycotoxin, produced by Fusarium nivale.[1],[2] Genotoxicity of nivalenol is evaluated by alkaline single-cell gel electrophoresis assay.[2]
Nivalenol is a secondary metabolite produced by Fusarium species, which is a plant pathogenic fungus of wheat and other cereals.[3]

Application

Nivalenol was used as standard to investigate in vitro mycotoxin binding of deoxynivalenol and nivalenol by adsorbent materials (activated carbon).[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nivalenol and deoxynivalenol by liquid chromatography/atmospheric pressure photoionization mass spectrometry
Tanaka H, et al.
Rapid Communications in Mass Spectrometry, 23(19), 3119- 3124 (2009)
Alaleh Zoghi et al.
Mini reviews in medicinal chemistry, 14(1), 84-98 (2013-12-18)
Removal of toxic metals and toxins using microbial biomass has been introduced as an inexpensive, new promising method on top of conventional methods for decontamination of food, raw material and concentrated. In this article the potential application of lactic acid
S Tsuda et al.
Mutation research, 415(3), 191-200 (1998-08-26)
We tested the genotoxicity of nivalenol (NIV), a potent toxic trichothecene from Fusarium nivale, in cultured CHO cells and in several mouse organs and tissues (liver, kidney, thymus, bone marrow and mucosa of stomach, jejunum, and colon) using the alkaline
Jungkwan Lee et al.
Applied and environmental microbiology, 78(7), 2161-2167 (2012-01-31)
Fusarium graminearum (Gibberella zeae) is an important pathogen of wheat, maize, barley, and rice in South Korea, and harvested grain often is contaminated with trichothecenes such as deoxynivalenol and nivalenol. In this study, we examined 568 isolates of F. graminearum
Tadahiro Suzuki et al.
Journal of agricultural and food chemistry, 60(37), 9519-9527 (2012-08-18)
Type B trichothecenes, deoxynivalenol (DON) and nivalenol (NIV), are secondary metabolites of Fusarium species and are major pollutants in food and feed products. Recently, the production trend of their derivatives, 3-acetyldeoxynivalenol (3-AcDON), 15-acetyldeoxynivalenol (15-AcDON), and 4-acetylnivalenol (4-AcNIV or fusarenon-X), has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service