Skip to Content
Merck
All Photos(2)

Key Documents

32997

Supelco

Sulfadoxine-d3

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H11N4O4S
CAS Number:
Molecular Weight:
313.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])Oc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC

InChI

1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)/i2D3

InChI key

PJSFRIWCGOHTNF-BMSJAHLVSA-N

General description

Sulfadoxine-d3 is an isotopically labeled compound of sulfonamide antibiotics. It is typically utilized as an analytical standard in chromatographic investigations.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfadoxine-d3 may be used as an analytical standard for the determination of several sulfonamides in:
  • Animal samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) and HPLC-MS/MS.
  • Wastewater samples by LC-MS/MS.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Confirmatory method for nine sulfonamides in miniature bovine muscle samples using HPLC/MS/MS without using SPE.
Nebot C, et al.
Journal of food and drug analysis, 18(3) (2010)
Determination of 19 sulfonamides residues in pork samples by combining QuEChERS with dispersive liquid-liquid microextraction followed by UHPLC-MS/MS.
Li X, et al.
Journal of Separation Science, 40(6), 1377-1384 (2017)
Analysis of multiclass antibiotic residues in urban wastewater in Tunisia.
Harrabi M, et al.
Environmental Nanotechnology, Monitoring & Management, 10, 163-170 (2018)
Simultaneous determination of sulfonamides, penicillins and coccidiostats in pork by high-performance liquid chromatography-tandem mass spectrometry.
Nebot C, et al.
Journal of Chromatographic Science, 50(5), 414-425 (2012)
Trace determination of sulfonamide antibiotics and their acetylated metabolites via SPE-LC-MS/MS in wastewater and insights from their occurrence in a municipal wastewater treatment plant.
Yuan SF, et al.
The Science of the Total Environment, 653(5), 815-821 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service