Skip to Content
Merck
All Photos(1)

Key Documents

46158

Supelco

Dapsone

VETRANAL®, analytical standard

Synonym(s):

4-Aminophenyl sulfone, 4,4′-Diaminodiphenyl sulfone, 4,4′-Sulfonyldianiline, Bis(4-aminophenyl) sulfone, DDS, Dapsone

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$95.20

$95.20


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$95.20

About This Item

Linear Formula:
(H2NC6H4)2SO2
CAS Number:
Molecular Weight:
248.30
Beilstein:
788055
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$95.20


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

175-177 °C (lit.)

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

InChI

1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

MQJKPEGWNLWLTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dapsone is a synthetic antibiotic belonging to the class of sulfonamides, which show inhibitory action against most gram-positive and many gram-negative organisms.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Dapsone may be used as an analytical reference standard for the quantification of the analyte in animal samples[1] and pharmaceutical formulations[2] using different analytical techniques
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 1B - STOT RE 2 - STOT SE 2

Target Organs

Blood, Blood,spleen,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A spectrophotometric method for the determination of metoclopramide HCl and dapsone
Revanasidadappa D.H and Manju B
Journal of Pharmaceutical and Biomedical Analysis, 25(3-4), 631-637 (2001)
C Curti et al.
European journal of medicinal chemistry, 42(6), 880-884 (2007-02-27)
An original one-pot microwave reaction was developed for the synthesis of sulfone derivatives as new potent antimicrobial agents. This eco-friendly methodology conducted in 30min led to desired products with good yields. The sulfones (4a and 4b) were obtained via the
F-R Zhang et al.
The New England journal of medicine, 369(17), 1620-1628 (2013-10-25)
Dapsone is used in the treatment of infections and inflammatory diseases. The dapsone hypersensitivity syndrome, which is associated with a reported mortality of 9.9%, develops in about 0.5 to 3.6% of persons treated with the drug. Currently, no tests are
Determination of sulfonamides in animal tissues using cation exchange reversed phase sorbent for sample cleanup and HPLC-DAD for detection.
Hela W, et al.
Food Chemistry, 83(4), 601-608 (2003)
Hakan M Gürcan et al.
American journal of clinical dermatology, 10(6), 383-396 (2009-10-15)
Dapsone is a chemotherapeutic agent primarily used in treating leprosy, Pneumocystis jiroveci (previously carinii) pneumonia, and malaria. It is also used as an adjuvant in the treatment of pemphigus and pemphigoid. To assess the role of dapsone in the treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service