Skip to Content
Merck
All Photos(1)

Key Documents

1.01541

Supelco

Bromocresol green sodium salt

pH indicator, water-soluble, ACS

Synonym(s):

Bromocresol green sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$686.00

$686.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$686.00

About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
MDL number:
UNSPSC Code:
12352107
EC Index Number:
263-657-4
NACRES:
NA.21

$686.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Bromocresol green sodium salt, indicator water-soluble ACS

grade

ACS reagent

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

loss

≤5.0% loss on drying, 110°C

visual transition interval

3.8-5.4, yellow-green to blue

storage temp.

15-25°C

SMILES string

[Na+].[S]1(=O)(=O)OC(c4c1cccc4)(c3c(c(c(c(c3)Br)[O-])Br)C)c2c(c(c(c(c2)Br)O)Br)C

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

Analysis Note

Transition range: pH 3.8 - pH 5.4yellow green - blue
Appearance of solution: conforms
Loss on drying (110 °C): ≤ 5.0 %
Transition range (according to ACS): conforms
ACS conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service