Skip to Content
Merck
All Photos(1)

Key Documents

07533

Millipore

Yeast Extract

suitable for microbiology

Synonym(s):

Yeast Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Product 07533 is not currently sold in your country. Contact Technical Service

biological source

Saccharomyces cerevisiae

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-N, ≥4.0%
nitrogen content, ≥8.0%

ign. residue

≤15%

loss

≤8.0% loss on drying

pH

5.8-6.8 (25 °C, 2% in H2O)

application(s)

food and beverages
microbiology

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

Application

A watersoluble yeast peptone enriched by the vitamin B complex for special fastidious microorganisms. Additionally it is enriched with Thiamin, Riboflavin, Niacin and Biotin. The yeast peptone contains highly concentrated protein in form of amino acids and peptides and serves as excellent source of minerals, nucleic acids and other promoters to support the growth of microorganisms.
Application: used as nutrient in diverse fermentations methods, including lactic acid cultures and pharma compounds.

Typical Analysis:

Viitamins mg/100 g product
Thiamin HCI (B1) ≥36
Riboflavin (B2) ≥13
Niacin ≥650
Vitamin B6 3.9
Folic acid 2.8
Pantothenic acid 11.2
Biotin ≥2
Inositol ≥160
Pyridoxine HCI 7

Minerals mg/100 g product
Calcium 164
Copper 0.8
Iron 0.7
Magnesium 144
Manganese 1.23
Phosphorus 1330
Potassium 2650
Sodium 678
Zinc 17.3

Analysis Note

solubility (2% in water, at 25 °C): clear to slightly turbid, light yellow and light brown-yellow and light brown

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How much Vitamin B12 is normally found in 100g of this product?

    1 answer
    1. The amount of Vitamin B12 in 100g of this product is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service