Skip to Content
Merck
All Photos(2)

Key Documents

1.17257

Millipore

Yeast Extract for Biotechnology and Microbiology

Prime, for Biotechnology and Microbiology, ≥4.8% Amino

Synonym(s):

Extract of Yeast, Yeast Extract

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$253.00
25 KG
$5,110.00

$253.00


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
1 KG
$253.00
25 KG
$5,110.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

$253.00


Estimated to ship on01 June 2025


Request more information

Product Name

Fermtech® Yeast Extract, For Biotechnology

Quality Level

Sterilization Compatibility

autoclavable compatible

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 25 kg

manufacturer/tradename

Fermtech®

storage condition

(Tightly closed)

technique(s)

microbiological culture: suitable

pH

6.7-7.3

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

General description

Fermtech® Yeast Extract is manufactured from selected strains of Saccharomyces cerevisiae. It is manufactured under controlled conditions to retain its vitamin content and other nutritive values, such as peptides, free amino acids, nucleic acids, minerals, trace elements, and phosphorous compounds. It is rich in vitamins and is often used to supply these factors in culture media at 0.3 - 0.5% concentration. It is produced through an auto-proteolytic reaction of Saccharomyces cells, resulting in a water-soluble peptone widely used as an ingredient of microbial culture media. Yeast extract provides a complete source of nutrients for microorganisms and is particularly useful for the cultivation of fastidious bacteria that require specific nutrients or additional growth factors. It can be used alone or in combination with other sources of nutrients, such as peptones or salts.

Application

Fermtech® Yeast Extract is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples. It is designed as a low endotoxin peptone suitable for use in fermentation for the cultivation of a broad range of fastidious microorganisms and cell culture media.

Legal Information

FERMTECH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service