Skip to Content
Merck
All Photos(1)

Key Documents

09182

Millipore

Yeast Extract

Synonym(s):

YE, Baker’s yeast extract

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$158.00
5 KG
$483.00

$158.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
$158.00
5 KG
$483.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040000
NACRES:
NA.85

$158.00


Available to ship on03 April 2025Details


Request a Bulk Order

biological source

Saccharomyces cerevisiae

Quality Level

sterility

non-sterile

Assay

≤3% amino-nitrogen basis
≥9% total nitrogen (N) basis

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≤3%
total nitrogen (N), ≥9%

packaging

pkg of 1 kg
pkg of 5 kg

impurities

≤16% ash
≤8% non-dissolved

loss

≤8% loss on drying

pH

6.5-7.5 (2% in H2O)

solubility

H2O: 2%, turbid, yellow

application(s)

food and beverages
microbiology

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Yeast extract is an extract of autolyzed yeast cells. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media. It is a non-animal and low endotoxin peptone suitable for use in fermentations media for cultivation and resuscitation of a large variety of fastidious microorganisms. It is rich in vitamins and other nutritive substances and so, recommended for general laboratory media

Application

Yeast extract is recommended for use in culture media and for the mass cultivation of various microorganisms. It provides multi-functional nutrient supplements that can also be used in microbial fermentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samriddha Ray et al.
The Journal of cell biology, 190(5), 793-805 (2010-09-02)
The mechanisms that regulate cytoskeletal remodeling during the transition between mitosis and interphase are poorly understood. In fission yeast the MOR pathway promotes actin polarization to cell tips in interphase, whereas the SIN signaling pathway drives actomyosin ring assembly and
Sara Hooshangi et al.
PLoS computational biology, 7(9), e1002172-e1002172 (2011-10-08)
Quorum sensing (QS) enables bacterial multicellularity and selective advantage for communicating populations. While genetic "switching" phenomena are a common feature, their mechanistic underpinnings have remained elusive. The interplay between circuit components and their regulation are intertwined and embedded. Observable phenotypes

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service