Skip to Content
Merck
All Photos(1)

Key Documents

40-4006

Sigma-Aldrich

Methyl Red − Methylene Blue solution

pH 5.4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000

form

liquid

availability

available only in Japan

pH

5.4

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service