Skip to Content
Merck
All Photos(1)

Key Documents

ZFPACKSP2

Milli-Q

Pack Support 2 Tube

with 2 gauges, For secure, leak-free fit of the pretreatment pack

Synonym(s):

Pack Support 2 Tube (with 2 gauges)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.13
Pricing and availability is not currently available. Contact Technical Service

packaging

pkg of 1 unit

compatibility

for use with AFS®

shipped in

ambient

General description

The Pack Support (2 tube) is a water purification accessory that comes with two pressure gauges. It provides a leak-free fit of replacement cartridge for MILLI-RO Compact Water Systems. The accessory is needed for installation and operation of PrePak 1 pretreatment pack.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLI-RO is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service