Skip to Content
Merck
All Photos(1)

Key Documents

YY2214265

Millipore

O-ring, 2-251, silicone

Accessories for filter holders for sample preparation.

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$273.00

$273.00


Estimated to ship on25 May 2025



Select a Size

Change View
4 EA
$273.00

About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

$273.00


Estimated to ship on25 May 2025


material

silicone O-ring

Quality Level

diam.

14.2 cm

compatibility

for use with Hazardous Waste System, 142mm

shipped in

ambient

General description

Accessories for filter holders for sample preparation.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service