Skip to Content
Merck
All Photos(1)

Key Documents

YT3009018-M

Millipore

Adapter, 1/8 NPTM to 1/4 tube

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$66.10

$66.10


Check Cart for Availability


Select a Size

Change View
1 EA
$66.10

About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

$66.10


Check Cart for Availability

material

316 stainless steel

Quality Level

fitting

1/4 in. O.D. inlet/outlet tube for 1/8 in. NPTM

compatibility

for use with ZHE Hazardous Waste Filtration System

shipped in

ambient

General description

Accessories for filter holders for sample preparation.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service