Key Documents
BAH195700000
SP Bel-Art® Tubing Connector Set
22 stepped and straight tubing connectors
Synonym(s):
connectors, tubing, tubing coupler
Select a Size
22 EACH
$130.00
$130.00
Check Cart for Availability
Select a Size
Change View
22 EACH
$130.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
$130.00
Check Cart for Availability
packaging
pack of 22 ea
manufacturer/tradename
Bel-Art H19570-0000
General description
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7mm (⅛ to ½").
Features and Benefits
  • Chemical-resistant polypropylene for use with a wide range of liquids or gases
  • Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors
  • FDA-grade polypropylene for direct contact with food, drugs, and cosmetics
  • Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)