Key Documents
XXSPTFF03
Millipore
Pellicon® Single-Pass TFF Cassette Retentate Collection Plate
About This Item
UNSPSC Code:
23151800
eCl@ss:
32031690
Pricing and availability is not currently available. Contact Technical Service
material
316L stainless steel housing
Quality Level
technique(s)
tangential flow filtration (TFF): suitable
fitting
3/4 in. sanitary flange drain/vent (TC)
application(s)
liquid formulation
General description
The Pellicon Single-Pass TFF Cassette Retentate Collection Plate is required to collect the retentate flow of single-pass TFF installations with Pellicon 3 0.57 m2, Pellicon 3 1.14 m2 or Pellicon 2 0.5 m2 cassettes.
Legal Information
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany