Skip to Content
Merck
All Photos(1)

Key Documents

XX2604701

Millipore

Filter holder support, PVC

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$350.00

$350.00


Available to ship on14 April 2025Details


Request more information

Select a Size

Change View
1 EA
$350.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$350.00


Available to ship on14 April 2025Details


Request more information

material

PVC body

Sterilization Compatibility

autoclavable compatible

input

sample type cosmetics
sample type beer
sample type pharmaceutical(s)
sample type bottled water

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

Application

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service