Skip to Content
Merck
All Photos(1)

Documents

STBMRFA94

Millipore

Fluid A

ready-to-use, bottle volume 900 mL , filling volume

Synonym(s):

Rinse Fluid, Washing Fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
eCl@ss:
32014001
NACRES:
NA.74

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

description

For rinsing and dilution

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

pack of 4 bottles
bottle of

bottle capacity

1000 mL

bottle volume

900 mL , filling volume

input

sample type pharmaceutical(s)

pH

7.1

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest®

shipped in

ambient

storage temp.

2-30°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Fluid A is a general rinse buffer in compliance with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. The product is a clear colorless liquid with a pH of 7.1 ± 0.2 at 25°C. It can be stored at 2-25°C (protected from light) for 12 months. Each box contains 4 bottles of 900 mL of fluid, with a screw cap with septum closure to reduce the risk of cross-contamination and optimize the disinfection procedures.

Application

Fluid A is used to rinse aqueous solutions during sterility testing by membrane filtration. It is used to dilute soluble solids for the same application. This product is also recommended as a rinsing fluid for membrane filtration of non-sterile products. It is excellent for dissolving or diluting samples, reconstituting commercial microorganisms, or as a transport medium for microorganisms.
Sterility testing
Bioburden testing

Components

Screw cap with septum

Culture Medium

Fluid A

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service