Skip to Content
Merck
All Photos(1)

Key Documents

PLCHK100205

Millipore

Ultracel® Ultrafiltration Membrane

Ultrafiltration, high recovery purification

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$898.00

$898.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 EA
$898.00

About This Item

UNSPSC Code:
41104903
eCl@ss:
32161003
NACRES:
NB.24

$898.00


Check Cart for Availability

Request a Bulk Order

material

composite regenerated cellulose (CRC) membrane

Quality Level

filtration area

20 cm2

membrane L × W

20 cm × 20 cm

matrix

Ultracel®

pore size

100 kDa NMWCO

application(s)

industrial qc (particle separation)

shipped in

ambient

General description

Device Configuration: Ultrafiltration Membrane
Filter Code: PLCHK

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service