Skip to Content
Merck
All Photos(1)

Key Documents

MXSMKFS48

Millipore

Milliflex® Pre-filled Media Cassettes

KF Streptococcus Agar, for fecal bacteria: Streptococcus spp., pack of 48 tests, suitable for bioburden testing

Synonym(s):

KF Streptococcus Agar Cassette

Sign Into View Organizational & Contract Pricing

Select a Size

48
$561.00

$561.00


Check Cart for Availability

Request more information

Select a Size

Change View
48
$561.00

About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

$561.00


Check Cart for Availability

Request more information

Agency

International standards

Quality Level

sterility

aseptically filled

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex Oasis®

color

clear purpleslightly opalescent

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

pH

7.2±0.2 (25.0 °C)

suitability

suitable for (suitable for solid cultivation)
fecal bacteria: Streptococcus spp.

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis®

shipped in

wet ice

storage temp.

2-8°C

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. KF Strep Agar is designed for the recovery of Enterococci (fecal streptococci) found in various types of water. Maltose and lactose serve as a source of energy and sodium azide is the selective agent. Bromocresol purple (indicator dye) turns to yellow when acid is produced and TTC is a redox indicator causing enterococci colonies to have a deep red colour as a result of tetrazolium reduction to an acidic azo dye.

Application

Milliflex® Cassette prefilled with KF Strep Agar is used in Bioburden testing and general microbiological analysis of water, wastewater and dairy products. It is widely used in pharmaceutical, cosmetic and brewing industry.

Specifications

United-States Pharmacopeia XXII Microbial Limits Tests

Culture Medium

KF Streptococcus

Analysis Note

QC organisms
S. faecalis (ATCC 29212)

E. coli (ATCC 8739) and E. aerogenes (ATCC 49701) as negative control
Streptococcal colonies appear red or pink surrounded by a yellow zone
Target Microorganisms: Fecal streptococci

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service