Skip to Content
Merck
All Photos(1)

Documents

1.46022

Millipore

HEIMPLATE MacConkey agar

pkg of (90 mm settle plate with 30 ml filling volume (20 or 120 plates per box)), For long incubation (LI), suitable for microbiology

Synonym(s):

MacConkey medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

EP 2.6.13
JP 4.05
USP 62

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

pkg of (90 mm settle plate with 30 ml filling volume (20 or 120 plates per box))

technique(s)

microbiological culture: suitable

pH

7.1 ( in H2O)

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Escherichia coli

General description

MacConkey Agar is a weakly differential medium for the selective growth of Enterobacteriaceae, specifically E. coli, from pharmaceutical products, food, water, and other materials. Enterobacteriaceae can be identified even in the presence of Salmonella and Shigella. Bile salts and crystal violet inhibit the growth of the Gram-positive microbial flora. Lactose and the pH indicator neutral red enable detection of lactose degradation. Bacteria degrading lactose to acids grow in pink to red-coloured colonies. Bacteria that cannot utilize lactose will grow colourless. Escherichia coli and other acid-forming bacteria will show a zone of precipitated bile salt around the colonies.The formulation of the medium (MacConkey Agar) is prepared as per recommendations of the current European, Japanese, and United States Pharmacopoeia and supplemented with neutralizers.

Application

MacConkey Agar - LI is designed for the isolation of Enterobacteriaceae, specifically E. coli, in pharmaceutical products, food, water, and other materials in controlled environments.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service