Skip to Content
Merck
All Photos(1)

Key Documents

MXSMCET48

Millipore

Milliflex® Pre-filled Media Cassettes

Cetrimide Agar, Naladixic Acid, for Pseudomonas aeruginosa, pack of 48 tests, suitable for bioburden testing

Synonym(s):

Cetrimide Agar Cassette

Sign Into View Organizational & Contract Pricing

Select a Size

48
$485.00

$485.00


Check Cart for Availability

Request more information

Select a Size

Change View
48
$485.00

About This Item

UNSPSC Code:
41116129
eCl@ss:
32160801
NACRES:
NA.74

$485.00


Check Cart for Availability

Request more information

Agency

International Standards

Quality Level

sterility

aseptically filled

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

composition

Naladixic Acid

packaging

cassette of 10 mL
pack of 48 tests

manufacturer/tradename

Milliflex Oasis®

color

light amberslightly opalescent

input

sample type beer
sample type bottled water
sample type cosmetics
sample type pharmaceutical(s)

pH

7.2±0.2 (25.0 °C)

suitability

suitable for (suitable for solid cultivation)
Pseudomonas aeruginosa

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. This medium is designed for the isolation and identification of Pseudomonas aeruginosa found in various types of water, especially high purity water. Cetrimide is the selective agent. It does not inhibit the pigment production of P. aeruginosa. In the presence of Cetrimide (which acts as a quaternary ammonium cationic detergent), bacterial cells growth other than P. aeruginosa is inhibited due to the release of nitrogen and phosphorous.

Application

Milliflex® Cassette prefilled with Cetrimide Agar is used in Bioburden testing and general microbiological analysis of water especially high purity water (electronics, RO, distilled or purified). It is widely used in pharmaceutical, cosmetic and brewing industry.

Specifications

United-States Pharmacopeia, European Pharmacopeia

Culture Medium

Cetrimide

Analysis Note

P. aeruginosa species appear as green to blue colonies with fluorescence under UV wavelength
QC organisms
P. aeruginosa (ATCC 9027)

P. mirabilis (ATCC 12453) as negative control
Target Microorganisms: Pseudomonas aeruginosa

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service