Skip to Content
Merck
All Photos(3)

Key Documents

MMGASKTQU

Millipore

MILLIFLEX® Quantum membrane transfer tool

for use with Milliflex® Quantum membrane transfer tool

Sign Into View Organizational & Contract Pricing

Select a Size

5 UNITS
$167.00

$167.00


Check Cart for Availability

Request more information

Select a Size

Change View
5 UNITS
$167.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

$167.00


Check Cart for Availability

Request more information

material

silicon

Quality Level

packaging

pack of 5 pieces (gaskets)

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex® Quantum membrane transfer tool

shipped in

ambient

Related Categories

General description

MILLIFLEX® Quantum system uses a fluorescent dye detection and can reduce your time to result by up to one third compared to traditional methods, enabling faster release or corrective actions. It works perfectly with the MILLIFLEX OASIS® system. The MILLIFLEX OASIS® system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance.
The MILLIFLEX® Quantum membrane transfer tool for MILLIFLEX OASIS® system is an accessory needed for dispensing the detection reagent on the membrane after filtration. This transfer tool contains spare gaskets. The MILLIFLEX® Quantum membrane transfer tool for MILLIFLEX OASIS® gaskets replacement kit contains 5 sets of gaskets enabling 5 replacements.

Application

The MILLIFLEX® Quantum and the MILLIFLEX OASIS® system finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service