BAF189790003
Poxygrid® Contact Plate and Petri Dish Rack
Holds 10 x 60 mm dishes, epoxy-coated (steel)
Synonym(s):
plate rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
epoxy-coated (steel)
feature
Holds 10 x 60 mm dishes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18979-0003
size
76x76x127 mm
compatibility
for use with Contact Plate and Petri Dishes
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Epoxy coated, steel wire rack is chemical and corrosion resistant and autoclavable at 121°C (250°F).
- Prevents accidents during transport
- Compact size maximizes incubator space and won′t crowd benchtop
Legal Information
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service