Skip to Content
Merck
All Photos(3)

Key Documents

MMGASKTEZ

Millipore

MILLIFLEX OASIS® filtration head gaskets replacement kit

for use with Milliflex Oasis® head for EZ-FIT® filtration units, pkg of 5 units (per pack)

Sign Into View Organizational & Contract Pricing

Select a Size

5 UNITS
$209.00

$209.00


Available to ship on07 April 2025Details


Request more information

Select a Size

Change View
5 UNITS
$209.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

$209.00


Available to ship on07 April 2025Details


Request more information

material

silicon

Quality Level

packaging

pkg of 5 units (per pack)

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® head for EZ-FIT® filtration units

shipped in

ambient

Related Categories

General description

Gasket spare part kit for Milliflex Oasis® head for EZ-Fit® Filtration unit is an additional accessory and allows to maintain the filtration head when requested. The EZ-Fit® filtration unit is a disposable filtration device for bioburden testing and have been designed to ensure optimal microbial recovery. The sterilized, disposable filtration devices streamline the workflow, save time and give highly reliable microbial enumeration results. Gasket kit for Milliflex Oasis® filtration head for EZ-Fit® filtration unit contains spare parts for five replacements.

Application

Gasket kit for Milliflex Oasis® Head for EZ-Fit® Filtration unit is an accessory to be used in Milliflex Oasis® filtration pump and finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical products analysis.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service