Skip to Content
Merck
All Photos(3)

Key Documents

MABT321

Sigma-Aldrich

Anti-Septin-9 Antibody, clone 1A6C2

clone 1A6C2, from rat

Synonym(s):

Septin-9, Eighth septin, Eseptin, Septin-like protein, SLP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A6C2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SEPTIN9(10801)

General description

Septin-9 (also known as eighth septin, Eseptin, and MLL septin-like fusion) is encoded by the Sept9 gene in rat and mouse (SEPT9 in human). Septins are GTP-binding proteins capable of forming higher-order oligomers and filamentous polymers that contact with the cell membrane and cytoskeleton. Septins are involved in various cellular processes, including cytokinesis, karyokinesis, exocytosis, and cell shape maintenance. The expression of SEPT2, SEPT6, SEPT7 and SEPT9 in heart tissue is reported to correlate well with cardiomyocyte cytokinesis activity during heart development, is highly expressed in embryonic heart, downregulated around birth, and nondetectable in the adult heart. (PMID 16739252 & 24837489). Clone 1A6C2 was generated using an immunogen sequence present only in rat spetin-9 isoform 1 (SLP-a; Q9QZR6), but missing in rat isoform 2-4 (SLP-b, E-septin long & short forms). The immunogen sequence is 100% conserved in murine species (A2A6U3) and differs by one amino acid when compared to the corresponding human sequence in isoform 2, and missing in human isoforms 1,3,4 (Q9UHD8).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of rat Septin-9.

Application

Anti-Septin-9 Antibody, clone 1A6C2 is an antibody against Septin-9 for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Septin-9 in HeLa cells (Dr. Taro Tachibana, Cell Engineering Corporation).
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Septin-9 in HeLa cell lysate (Dr. Taro Tachibana, Cell Engineering Corporation).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Septin-9 in 10 µg of NIH/3T3 cell lysate.

Target description

~65 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service