Key Documents
BR475800
BRAND® slide box
25 slides, 76 x 26 mm
Synonym(s):
Microscope slides, Microscopy, Slide box, Staining, Staining Accessories
Select a Size
1 EA
$28.80
$28.80
Available to ship on28 April 2025Details
Select a Size
Change View
1 EA
$28.80
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
$28.80
Available to ship on28 April 2025Details
material
polystyrene
feature
Holds 25 x slides 76x26 mm
packaging
pack of 1 ea
manufacturer/tradename
BRAND 475800
General description
Sturdy design. Screw lid with dealing wire lug. Ideal for storing or shipping 5 thick or 10 thin slides. Slides protrude 10 mm above the box, to facilitate removal.
For slides, size (in, mm) 76 x 26. Numbered slots in base. Lid with index card.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)