Skip to Content
Merck
All Photos(1)

Key Documents

IFX-PVER-K20

Millipore

xMAP INTELLIFLEX Ver Kit, 20 Uses, RUO

Synonym(s):

Multiplex assay kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$934.00

$934.00


Available to ship on14 April 2025Details



Select a Size

Change View
1 EACH
$934.00

About This Item

UNSPSC Code:
41116010
NACRES:
NC.54

$934.00


Available to ship on14 April 2025Details


packaging

kit of 1

compatibility

for use with INTELLIFLEX RUO System and INTELLIFLEX-DRSE

storage temp.

2-8°C

General description

For importing target values from the xMAP® INTELLIFLEX Calibration and Performance Verification Kits

Application

Multiplexing

Components

  • Luminex Classification Control Microspheres
  • MagPlex Classification Control Microspheres
  • Luminex Reporter Control Microspheres
  • xMAP Fluidics 1 Microspheres
  • xMAP Fluidics 2 Microspheres
  • Performance Verification CD
  • 28 strip wells

Legal Information

xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Get even more out of your samples with this Luminex® technology-based mouse cytokine assay – the MILLIPLEX® Mouse Cytokine Expanded Panel 1.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service