Skip to Content
Merck
All Photos(2)

Documents

40-012

Millipore

Luminex 200 System

The Luminex 200 system is a compact analyzer that performs up to 100 immunoassays simultaneously using a single drop of fluid, therefore requiring very small samples when compared with other testing technologies.

Synonym(s):

Luminex Multiplexing Platform, Luminex xmap Instrument, Millipore Multiplexing Instrument

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32161000
NACRES:
NA.32

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric

shipped in

ambient

General description

The Luminex 200 system, a compact analyzer, can perform up to 100 immunoassays using a single drop of fluid simultaneously. This system employs a 3.1 ”Xponent” version of the xMAP® technology operating system. The Luminex 200system used with MILLIPLEX® map magnetic bead-based multi-analyte panels, MILLIPLEX® analysis 5.1 software, and technical support, offers a broad mid to high range solution for fast and precise quantification of biomarkers.

Application

Luminex 200 System is suitable for multiplexing.

Features and Benefits

  • Flexible to run magnetic and non-magnetic bead assays
  • Up to 100 analytes per well of a 96- well plate in as little as 25 μL of the sample
  • Easy to use
  • Scalable due to bar code reader addition

Linkage

Replaces: 40-01

Legal Information

Luminex 200 system is a trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service