Skip to Content
Merck
All Photos(1)

Key Documents

40-50030

Millipore

MAGPIX® Drive Fluid PLUS 4PK

Synonym(s):

Drive Fluid, Luminex, MAGPIX Drive Fluid PLUS 4PK, MPXDF4PK, magpix

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$133.00

$133.00


Available to ship on08 April 2025Details



Select a Size

Change View
1 EACH
$133.00

About This Item

UNSPSC Code:
41116010
NACRES:
NA.25

$133.00


Available to ship on08 April 2025Details


form

liquid

packaging

pkg of 4 pack, 700 mL each

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

storage temp.

room temp

General description

N/A

Application

N/A

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MAGPIX is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Need help deciding which multiplex assay fits your research? Before you start multiplexing with MILLIPLEX® multiplex kits, check out these tips for how to decide what the best MILLIPLEX® assay is for your research. Also, learn what materials are required to complete multiplex workflows but not provided with the kits.

The newer xMAP® Sheath Fluid PLUS formulation contains 3 modes of microbial action to eliminate potential environmental microbial resistance. See how the preparations compare in this performance comparison of the previous xMAP® Sheath Fluid to the new xMAP® Sheath Fluid PLUS formulation in MILLIPLEX® multiplex assays.

Questions

  1. What are the distinctions between the Magpix Drive Fluid and Drive Fluid Plus? Additionally, how can one transition to Drive Fluid Plus on the MagPix?

    1 answer
    1. The Drive Fluid formulation is being updated from a single preservative to a mixture of three preservatives. This change is intended to enhance transit stability and reduce the likelihood of future acquired microbial resistance.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service