ACK5100PG
Amicon® Pro Affinity Concentration Kit Protein G with 100kDa Amicon® Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying & concentrating Antibodies.
About This Item
Recommended Products
material
Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)
sterility
non-sterile
product line
Amicon®
manufacturer/tradename
Amicon® Pro
parameter
10 mL sample volume
technique(s)
buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)
L
12 cm (4.7 in.)
diam.
2.97 cm
filtration area
1 cm2
pore size
100 kDa NMWCO
shipped in
wet ice
General description
Application
Features and Benefits
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Met. Corr. 1
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service