Skip to Content
Merck
All Photos(2)

Key Documents

ABN1452

Sigma-Aldrich

Anti-TRIO

from rabbit, purified by affinity chromatography

Synonym(s):

Triple functional domain protein, EC: 2.7.11.1.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TRIO(7204)

General description

Triple functional domain protein (UniProt: O75962; also known as EC: 2.7.11.1, PTPRF-interacting protein, TRIO) is encoded by the TRIO gene (Gene ID: 7204) in human. TRIO is a complex protein that is widely expressed in human tissue, with highest levels reported in heart, skeletal muscle, and brain. Five isoforms of TRIO have been reported that are produced by alternative splicing. It contains three catalytic domains, two tandem Dbl Homology (DH) and pleckstrin homology (PH) domains, and a serine/Threonine kinase domain. The DH-PH module is involved in guanine nucleotide exchange and the activation of the small GTPases of the Rho family. The DH1-PH1 domain (GEF1) is reported to activate the Rho G and Rac1, whereas DH2-PH2 (GEF2) is involved in the activation of RhoA. TRIO is involved in coordinating actin remodeling, which is necessary for cell migration and growth. In developing hippocampal neurons it limits dendrite formation, without affecting the establishment of axon polarity. After dendrites are formed, it is reported to be involved in the control of synaptic function by regulating the endocytosis of AMPA-selective glutamate receptors at CA1 excitatory synapses. Mutations in TRIO gene have been linked to mental retardation and microcephaly. (Ref.: Debant, A. et al. (1996). Proc. Natl. Acad. Sci, USA. 93(11); 5466-71; Bellanger, JM. Et al. (2000). Nat. Cell Biol. 2(12); 888-92).

Specificity

This rabbit polyclonal antibody detects human Triple functional domain protein (TRIO). It targets an epitope within 17 amino acids from the C-terminal half. The immunogen sequence is conserved in all 5 isoforms.

Immunogen

KLH-conjugated liner peptide corresponding to 17 amino acids from the C-terminal half of human Triple functional domain protein.

Application

Anti-TRIO Antibody, Cat. No. ABN1452, is a highly specific rabbit polyclonal antibody that targets Triple functional domain protein (TRIO) and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected TRIO in human cerebral cortex, human pancreas, and human testis tissue.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TRIO in 10 µg of HEK293 cell lysate.

Target description

~300 kDa observed; 346.90 kDa calculalted. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service