Skip to Content
Merck
All Photos(2)

Key Documents

HPA001869

Sigma-Aldrich

ANTI-FAT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cadherin-related tumor suppressor homolog precursor, Anti-FAT, Anti-Protein fat homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAT1(2195)

Immunogen

Cadherin-related tumor suppressor homolog precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FAT antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
The antibody has been used for sandwich ELISA to determine levels of soluble Fat1 in pancreatic cancer serum samples.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

FAT1 (FAT atypical cadherin 1), transmembrane protein belonging to the cadherin super family, encodes a large transmembrane protein with 34 tandem cadherin repeats, five EGF (epidermal growth factor)-like repeats, and a laminin A-G domain. It exhibits tumor suppressor activity. It is ubiquitously expressed in epithelial tissues. Specifically, FAT1 is localized to the cell membrane, often concentrated at filopodia, lamellipodia, and sites of cell-cell contact. In mammals, four isoforms have been found: FAT1, FAT2, FAT3 and FAT4. FAT1 helps to regulate cell-cell association and actin dynamics.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85144

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Dunne et al.
Genomics, 30(2), 207-223 (1995-11-20)
FAT, a new member of the human cadherin super-family, has been isolated from the T-leukemia cell line J6. The predicted protein closely resembles the Drosophila tumor suppressor fat, which is essential for controlling cell proliferation during Drosophila development. The gene
Xiaoling Hu et al.
Oncology reports, 39(5), 2136-2146 (2018-03-23)
FAT atypical cadherin 1 (FAT1) belongs to the cadherin superfamily and has been reported to regulate cell‑cell adhesion and other cell behaviors, suggesting its pivotal roles in human cancers. We previously identified FAT1 as one of the significant mutant genes in esophageal
Zhiqiang Li et al.
Cancer cell, 34(6), 893-905 (2018-12-12)
Cyclin dependent kinase 4/6 (CDK4/6) inhibitors (CDK4/6i) are effective in breast cancer; however, drug resistance is frequently encountered and poorly understood. We conducted a genomic analysis of 348 estrogen receptor-positive (ER+) breast cancers treated with CDK4/6i and identified loss-of-function mutations
Anne E Roehrig et al.
PloS one, 16(8), e0254697-e0254697 (2021-08-24)
The PAF complex (PAFC) coordinates transcription elongation and mRNA processing and its CDC73/parafibromin subunit functions as a tumour suppressor. The NF2/Merlin tumour suppressor functions both at the cell cortex and nucleus and is a key mediator of contact inhibition but
Nathalie Wojtalewicz et al.
PloS one, 9(3), e90461-e90461 (2014-03-15)
In pancreatic cancer, there is a clear unmet need to identify new serum markers for either early diagnosis, therapeutic stratification or patient monitoring. Proteomic analysis of tumor cell secretomes is a promising approach to indicate proteins released from tumor cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service