Skip to Content
Merck
All Photos(1)

Key Documents

BAF451000001

Scienceware® gel incubating tray

W × L 9.0 cm (3.5 in.) × 9.0 cm (3.5 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$79.00

$79.00


Check Cart for Availability


Select a Size

Change View
5 EA
$79.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

$79.00


Check Cart for Availability

packaging

pack of 5 ea

manufacturer/tradename

Bel-Art F45100-0001

W × L

9.0 cm (3.5 in.) × 9.0 cm (3.5 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Minimize the use of expensive antibodies when incubating filters in the Western Blot process by using an optimally sized tray.

  • Includes attached lid to protect against spillage
  • Trays are stackable, reclosable and reusable
  • Molded of polypropylene
  • Autoclavable at 121°C (250°F)

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jose R Quejada et al.
Nucleic acids research, 45(20), e172-e172 (2017-10-19)
Light-inducible systems allow spatiotemporal control of a variety of biological activities. Here, we report newly optimized optogenetic tools to induce transcription with light in mammalian cells, using the Arabidopsis photoreceptor Flavin Kelch-repeat F-box 1 (FKF1) and its binding partner GIGANTEA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service