Skip to Content
Merck
All Photos(2)

Key Documents

ABE1027

Sigma-Aldrich

Anti-NUBP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cytosolic Fe-S cluster assembly factor NUBP1, Nucleotide-binding protein 1, NBP 1, NUBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NUBP1(4682)

General description

NUBP1 also known as NBP as is expressed in several tissues with the highest being in the lung and testis. NUBP1 appears to interact with actin and the cytoskeleton and is required for centrosome duplication but it is also need for proper synthesis of iron sulfur proteins and the protein also binds ATP. NUBP1 has roles in cell growth, cellular iron homeostasis, centrosome localization, iron-sulfur cluster assembly, and apolipoprotein modification. NUBP1 is localized to the cytoplasm. Interestingly too, recent research indicates that mutations in NUBP1 in mice lead defects in lung development and eye cataracts, many of which are traced back to NUBP1 role in regulating centrosome dynamics and microtubule organization.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human NUBP1.

Application

Anti-NUBP1 Antibody | ABE1027 is an antibody against NUBP1 Antibody for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected NUBP1 in HEK293 and HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected NUBP1 in 10 µg of HeLa cell lysate.

Target description

~35 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service