Skip to Content
Merck
All Photos(2)

Key Documents

ABS1507

Sigma-Aldrich

Anti-Aurora B

from rabbit, purified by affinity chromatography

Synonym(s):

Aurora kinase B, EC:2.7.11.1, Aurora 1, Aurora- and IPL1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, ARK-2, Aurora-related kinase 2, STK-1, Serine/threonine-protein kinase 12, Serine/threonine-protein kinase 5, Serine/threonin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$452.00

$452.00


Estimated to ship on18 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1149

Request a Bulk Order

Select a Size

Change View
100 μG
$452.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$452.00


Estimated to ship on18 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1149

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... AURKB(9212)
mouse ... Aurkb(20877)

General description

Aurora kinase B (UniProt: Q96GD4; also known as EC:2.7.11.1, Aurora 1, Aurora- and IPL1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, ARK-2, Aurora-related kinase 2, STK-1, Serine/threonine-protein kinase 12, Serine/threonine-protein kinase 5, Serine/threonine-protein kinase aurora-B) is encoded by the AURKB (also known as AIK2, AIM1, AIRK2, ARK2, STK1, STK12, STK5) gene (Gene ID: 9212) in human. Aurora Kinase B is a serine/threonine-protein kinase component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. It is a key component of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Aurora kinase B phosphorylates the CPC complex subunits BIRC5/survivin, CDCA8/borealin and INCENP and phosphorylation of INCENP leads to increased Aurora kinase B activity. Five isoforms of Aurora Kinase are reported that are produced by alternative splicing. Its expression is shown to be cell-cycle regulated, with low expression in G1/S phase and an increase during G2 and M phase. Following M phase its expression declines. Cancer cells display high expression of Aurora kinase B during M phase. Disruption in regulation of Aurora kinase B causes perturbation of chromosomal integrity in cancer cells through its dominant-negative effect on cytokinesis. High level expression is observed in the thymus. It is also expressed in the spleen, lung, testis, colon, placenta and fetal liver. Substitution of Lysine 106 with arginine leads to loss of its kinase activity and severely impairs mitotic progression.

Specificity

This rabbit polyclonal antibdy detects Aurora kinase B in human cells.

Immunogen

Epitope: C-terminus
KLH-conjugated Linear Peptide corresponding to the 11 amino acids from the C-terminal region of Human Aurora kinase B.

Application

Anti-Aurora B, Cat. No. ABS1507, is a rabbit polyclonal antibody that detects Aurora Kinase B and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Aurora B in HeLa cells.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Aurora B in 10 µg of HeLa cell lysate.

Target description

~39 kDa observed; 39.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service