Key Documents
Z682853
Ace receiving flask,
capacity 500 mL, joint: ST/NS 35/20, plastic-coated glass
Select a Size
1 EA
$190.00
$190.00
Check Cart for Availability
Select a Size
Change View
1 EA
$190.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$190.00
Check Cart for Availability
material
plastic-coated glass
round bottom flask
single-neck
Joint
joint: ST/NS35/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Ace Glass 3996-08
capacity
500 mL
General description
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.
Choose from one of the most recent versions:
Certificates of Analysis (COA)