Skip to Content
Merck
All Photos(2)

Key Documents

AB3503-I

Sigma-Aldrich

Anti-PDX-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pancreas/duodenum homeobox protein 1, PDX-1, Glucose-sensitive factor, GSF, Insulin promoter factor 1, IPF-1, Insulin upstream factor 1, IUF-1, Islet/duodenum homeobox-1, IDX-1, Somatostatin-transactivating factor 1, STF-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$471.00

$471.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$471.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$471.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDX1(3651)

General description

Pancreatic and duodenal homeobox 1, also known as PDX1, and Insulin promoter factor 1 is a human gene. This islet-specific protein activates transcription of the insulin andsomatostatin genes. It is a key regulator of islet peptide hormone expressionn and also plays an essential role in pancreatic development. Mutations in this gene may be involved in several disorders of the pancreas or in diabetes mellitus.

Specificity

Recognizes PDX-1

Immunogen

KLH-conjugated linear peptide corresponding to region amino acids 90-120 of human PDX-1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected pdx-1 in the islets of Langerhans of human and mouse pancreas.
This Anti-PDX-1 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of PDX-1.

Quality

Evaluated by Western Blotting in mouse pancreatic beta cell tissue lysate.

Western Blotting Analysis: A 1:4,000 dilution of this antibody detected PDX-1 in mouse pancreatic beta cell tissue lysate.

Target description

~38 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service