Skip to Content
Merck
All Photos(1)

Key Documents

344932

Sigma-Aldrich

Furin, Human, Recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$936.00

$936.00


Estimated to ship on28 May 2025



Select a Size

Change View
50 μL
$936.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$936.00


Estimated to ship on28 May 2025


form

liquid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

Related Categories

General description

Recombinant, human furin expressed in vaccinia virus (VV:hFUR713t) infected BSC-40 cells. Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like pro-hormone convertase (PC) family. Members of this family cleave pro-hormones, neuropeptide precursors, some viral proteins, and bacterial toxins. The preparation was subjected to UV irradiation for 5 minutes to inactivate any residual virus.
This preparation was subjected to UV irradiation for 5 min to inactivate any residual vaccinia virus.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of furin that will cleave 1.0 pmol AMC from the substrate pGlu-Arg-Thr-Lys-Arg-AMC per min at 30°C, pH 7.5.

Physical form

In 20 mM Bis-Tris buffer, 1 mg/ml BSA, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Bassi, D.E., et al. 2001. Mol. Carcinog.31, 224.
Bassi, D.E., et al. 2001. Proc. Natl. Acad. Sci. USA98, 10326.
Creemers, J.W., et al. 2001. J. Biol. Chem.276, 4211.
Wang, X., and Pei, D. 2001. J. Biol. Chem.276, 35953.
Molloy, S.S., et al. 1992. J. Biol. Chem.267, 16396.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service