Skip to Content
Merck
All Photos(1)

Key Documents

901781

Supelco

Seal (O-ring 14x2.5 mm)

for use with withdrawl systems (products 1.01114 and 1.01123)

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$117.00

$117.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
$117.00

About This Item

UNSPSC Code:
41121516

$117.00


Estimated to ship on28 May 2025


Request a Bulk Order

compatibility

for use with withdrawl systems (products 1.01114 and 1.01123)

shipped in

ambient

storage temp.

room temp

General description

If withdrawal system parts are lost or damaged, spare parts are available, e.g. special sealings for our withdrawal systems. This seal fits to the manual withdrawal systems 1.01114.0001 and 1.01123.0001.
for withdrawal systems Cat. Nos. 1.01114.0001 and 1.01123.0001

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service