Skip to Content
Merck
All Photos(1)

Key Documents

8.52045

Sigma-Aldrich

Fmoc-Gln(Trt)-OH

Novabiochem®

Synonym(s):

Fmoc-Gln(Trt)-OH, N-α-Fmoc-N-γ-trityl-L-glutamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$112.00
100 G
$337.00
250 G
$715.00
500 G
$1,350.00
1 KG
$2,490.00
2 KG
$4,800.00
5 KG
$9,360.00

$112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$112.00
100 G
$337.00
250 G
$715.00
500 G
$1,350.00
1 KG
$2,490.00
2 KG
$4,800.00
5 KG
$9,360.00

About This Item

Empirical Formula (Hill Notation):
C39H34N2O5
CAS Number:
Molecular Weight:
610.70
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥93.0% (acidimetric)
≥98% (TLC)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

110-125 °C

application(s)

peptide synthesis

functional group

amide

storage temp.

-10 to -25°C

SMILES string

N([C@@H](CCC(=O)NC(c6ccccc6)(c5ccccc5)c4ccccc4)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C39H34N2O5/c42-36(41-39(27-14-4-1-5-15-27,28-16-6-2-7-17-28)29-18-8-3-9-19-29)25-24-35(37(43)44)40-38(45)46-26-34-32-22-12-10-20-30(32)31-21-11-13-23-33(31)34/h1-23,34-35H,24-26H2,(H,40,45)(H,41,42)(H,43,44)/t35-/m0/s1

InChI key

WDGICUODAOGOMO-DHUJRADRSA-N

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Fmoc-Gln(Trt)-OH has good solubility properties in most organic solvents, and its use has been shown to result in significantly purer peptides than other derivatives used for the introduction of Gln [1]. Coupling can be performed by standard procedures. The trityl-protecting group is normally removed by 95% TFA in 1-3 hours, with no alkylation of Trp.

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] P. Sieber, et al. (1991) Tetrahedron Lett., 32, 739

Application

  • Substrate specificity profiling of SARS-CoV-2 Mpro protease provides basis for anti-COVID-19 drug design: This study highlights the use of Fmoc-Gln(Trt)-OH in the synthesis of peptide substrates for biochemical assays, contributing to the development of COVID-19 therapeutics (Rut et al., 2020).

Linkage

Replaces: 04-12-1090

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 160 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Gln (Trt) -OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Gln(Trt)-Gln(Trt)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Gln-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 93.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the

Related Content

Purer Fmocs Means Purer Peptides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service