Skip to Content
Merck
All Photos(1)

Key Documents

8.52044

Sigma-Aldrich

FMOC-Asn(Trt)-OH

N-α-Fmoc-N-ß-trityl-L-asparagine Novabiochem®

Synonym(s):

FMOC-Asn(Trt)-OH, N-α-Fmoc-N-β-trityl-L-asparagine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$112.00
100 G
$336.00
250 G
$711.00
500 G
$1,350.00
1 KG
$2,480.00
2 KG
$4,800.00
5 KG
$9,360.00

$112.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
25 G
$112.00
100 G
$336.00
250 G
$711.00
500 G
$1,350.00
1 KG
$2,480.00
2 KG
$4,800.00
5 KG
$9,360.00

About This Item

Empirical Formula (Hill Notation):
C38H32N2O5
CAS Number:
Molecular Weight:
596.67
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

$112.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥98.0% (acidimetric)
≥99.0% (HPLC)

form

powder

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

210-220 °C

solubility

<0.00005 g/L

application(s)

peptide synthesis

functional group

amide

storage temp.

−20°C (−15°C to −25°C)

SMILES string

N([C@@H](CC(=O)NC(c6ccccc6)(c5ccccc5)c4ccccc4)C(=O)[O-])C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C38H32N2O5/c41-35(40-38(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28)24-34(36(42)43)39-37(44)45-25-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33/h1-23,33-34H,24-25H2,(H,39,44)(H,40,41)(H,42,43)/p-1/t34-/m0/s1

InChI key

KJYAFJQCGPUXJY-UMSFTDKQSA-M

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Fmoc-Asn(Trt)-OH has good solubility properties in most organic solvents, and its use has been shown to result in significantly purer peptides than other derivatives used for the introduction of Asn [1,2]. Coupling can be performed by standard procedures. The trityl group is normally removed by 95% TFA in 1-3 hours, with no alkylation of Trp residues. When Asn(Trt) is the N-terminal residue, the reaction time may need to be extended to ensure complete deprotection [3].

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] P. Sieber, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 577.
[2] P. Sieber, et al. (1991) Tetrahedron Lett., 32, 739.
[3] M. Friede, et al. (1992) Pept. Res., 5, 145.

Application

  • Synthesis of a burkholdine analogue containing β-hydroxytyrosine: This study discusses the synthesis of a burkholdine analogue, utilizing Fmoc-Asn(Trt)-OH as part of the solid support loading process (Kato et al., 2019).

Linkage

Replaces: 04-12-1089

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.7 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Asn (Trt)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Asn(Trt)-Asn(Trt)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Asn-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.05 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Purer Fmocs Means Purer Peptides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service