Skip to Content
Merck
All Photos(1)

Key Documents

8.52001

Sigma-Aldrich

Fmoc-Gly-OH

Novabiochem®

Synonym(s):

Fmoc-Gly-OH, FMOC-Glycine, N-α-Fmoc-glycine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$59.10
100 G
$167.00
250 G
$333.00
500 G
$500.00
1 KG
$919.00
2 KG
$1,810.00
5 KG
$4,440.00

$59.10


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 G
$59.10
100 G
$167.00
250 G
$333.00
500 G
$500.00
1 KG
$919.00
2 KG
$1,810.00
5 KG
$4,440.00

About This Item

Empirical Formula (Hill Notation):
C17H15NO4
CAS Number:
Molecular Weight:
297.31
MDL number:
UNSPSC Code:
12352209
EC Index Number:
249-373-3
NACRES:
NA.22

$59.10


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥98.0% (acidimetric)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

177.4 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N(CC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C17H15NO4/c19-16(20)9-18-17(21)22-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15H,9-10H2,(H,18,21)(H,19,20)

InChI key

NDKDFTQNXLHCGO-UHFFFAOYSA-N

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of glycine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-12-1001

Analysis Note

Color (visual): white to off white
Appearance of substance (visual): powder
Color index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Gly-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Gly-Gly-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service