Skip to Content
Merck
All Photos(1)

Key Documents

8.41643

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzylbromide

for synthesis

Synonym(s):

2,3,4,5,6-Pentafluorobenzylbromide, Bromomethylpentafluorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$281.00

$281.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$281.00

About This Item

Empirical Formula (Hill Notation):
C7H2BrF5
CAS Number:
Molecular Weight:
260.99
MDL number:
UNSPSC Code:
12352101
EC Index Number:
217-182-4
NACRES:
NA.22

$281.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

transition temp

flash point 83 °C

density

1.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Fc1c(c(c(c(c1F)F)CBr)F)F

InChI

1S/C7H2BrF5/c8-1-2-3(9)5(11)7(13)6(12)4(2)10/h1H2

InChI key

XDEPVFFKOVDUNO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.845 - 1.855
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service