Skip to Content
Merck
All Photos(1)

Key Documents

263397

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzyl bromide

97%

Synonym(s):

1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$39.20
5 G
$93.30

$39.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$39.20
5 G
$93.30

About This Item

Linear Formula:
(CF3)2C6H3CH2Br
CAS Number:
Molecular Weight:
307.03
Beilstein:
656506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$39.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.445 (lit.)

density

1.675 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1cc(CBr)cc(c1)C(F)(F)F

InChI

1S/C9H5BrF6/c10-4-5-1-6(8(11,12)13)3-7(2-5)9(14,15)16/h1-3H,4H2

InChI key

ATLQGZVLWOURFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)benzyl bromide has been used:
  • as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry[1]
  • in enantioselective synthesis of non-peptidic neurokinin NK1 receptor antagonist, L-733,060[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of L-733,060.
Bhaskar G and Venkateswara Rao B.
Tetrahedron Letters, 44(5), 915-917 (2003)
B C Blount et al.
Analytical biochemistry, 219(2), 195-200 (1994-06-01)
A sensitive method using gas chromatography and negative chemical ionization mass spectrometry for the detection of uracil in DNA is described. Uracil DNA glycosylase is used to specifically cleave uracil from DNA. Once removed, uracil is derivatized with 3,5-bis(trifluoromethyl)benzyl bromide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service