Skip to Content
Merck
All Photos(1)

Key Documents

70480

Supelco

α-Naphtholbenzein

indicator (pH 8.2-10.0)

Synonym(s):

4,4′-(α-Hydroxybenzylidene)di-1-naphthol, p-naphtholbenzein

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$182.00
50 G
$507.00

$182.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$182.00
50 G
$507.00

About This Item

Empirical Formula (Hill Notation):
C27H18O2
CAS Number:
Molecular Weight:
374.43
Beilstein:
2628063
EC Number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

$182.00


Available to ship on03 April 2025Details


Request a Bulk Order

grade

indicator (pH 8.2-10.0)

Quality Level

form

powder

technique(s)

UV/Vis spectroscopy: suitable

mp

230-235 °C (lit.)

SMILES string

Oc1ccc(\C(c2ccccc2)=C3\C=CC(=O)c4ccccc34)c5ccccc15

InChI

1S/C27H18O2/c28-25-16-14-23(19-10-4-6-12-21(19)25)27(18-8-2-1-3-9-18)24-15-17-26(29)22-13-7-5-11-20(22)24/h1-17,28H/b27-24-

InChI key

VDDWRTZCUJCDJM-PNHLSOANSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Naphtholbenzein is a dye generally used during acid-base titration method.[1]

Application

α-Naphtholbenzein has been used as indicator during sequential injection analysis technique done for acid-base titrations.[1]

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titration of strong and weak acids by sequential injection analysis technique.
Maskula, Silla, Johan Nyman, and Ari Ivaska
Talanta, 52.1, 91-99 (2000)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service