Skip to Content
Merck
All Photos(1)

Documents

8.40124

Sigma-Aldrich

Span® 85

for synthesis

Synonym(s):

Span® 85, Sorbitan trioleate, (Z,Z,Z)-Sorbitan tri-9-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H108O8
CAS Number:
Molecular Weight:
957.49
MDL number:
UNSPSC Code:
12352108
EC Index Number:
247-569-3
NACRES:
NA.22

vapor pressure

<1.4 hPa ( 20 °C)

Quality Level

form

liquid

potency

39800 mg/kg LD50, oral (Rat)

kinematic viscosity

120 cSt(40 °C)

transition temp

flash point 225 °C

solubility

insoluble <0.001 g/L

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C60H108O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-53-55(67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2)60-59(54(61)52-66-60)68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3/h25-30,54-55,59-61H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-/t54-,55?,59+,60-/m1/s1

InChI key

PRXRUNOAOLTIEF-XDTJCZEISA-N

Application


  • Macromolecular Stabilization: Span® 85 is used as a stabilizer for the preparation of PMMA colloidal particles in nonpolar media, proving effective in maintaining stability under various conditions, crucial for industrial applications in coatings and advanced materials manufacturing (Liu et al., 2022).

  • Enhanced Nanoemulsion Systems: It plays a critical role in the formation of aerosolized nanoemulsion systems encapsulating docetaxel and curcumin, utilized in cancer therapy to improve therapeutic outcomes through enhanced drug delivery mechanisms (Abdul Rahman et al., 2020).

  • Skin Protection Formulations: Span® 85 is integral in the formulation and clinical assessment of microemulsion-based hydrogels for UV protection, demonstrating its utility in skincare products by enhancing sun protection factor and skin compatibility (Ebrahimi et al., 2019).

  • Transdermal Drug Delivery: This surfactant is utilized in tailoring novel soft nano-vesicles, known as ′Flexosomes′, to enhance transdermal drug delivery, which is pivotal for non-invasive therapeutic applications and improving patient compliance (Mansour et al., 2019).

Analysis Note

Hydroxyl value: 55 - 75
Saponification value: 170 - 190
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

437.0 °F

Flash Point(C)

225 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service